Assay Details
Gene Symbol: LOC5568117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652006.2 | XP_001652056.2 | 1 | 57 | ||
GenBank mRNA | M79452.1 | - | 1 | 1014 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
