Assay Details
Gene Symbol: LOC5565800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650117.2 | XP_001650167.2 | 6 | 2325 | 112 | |
XM_021849525.1 | XP_021705217.1 | 7 | 3019 | 112 | ||
XM_021849526.1 | XP_021705218.1 | 7 | 3063 | 112 | ||
XM_021849527.1 | XP_021705219.1 | 7 | 2831 | 112 | ||
XM_021849528.1 | XP_021705220.1 | 7 | 2584 | 112 | ||
XM_021849529.1 | XP_021705221.1 | 7 | 2647 | 112 | ||
XM_021849530.1 | XP_021705222.1 | 7 | 2390 | 112 | ||
GenBank mRNA | AY433395.1 | - | 1 | 422 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
