Assay Details
Gene Symbol: LOC5576381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844033.1 | XP_021699725.1 | 5 | 2420 | 86 | |
GenBank mRNA | AY433417.1 | - | 1 | 323 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
