Assay Details
Gene Symbol: LOC5571385 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839279.1 | XP_021694971.1 | 1 - 2 | 350 | 116 | |
GenBank mRNA | AY433445.1 | - | 1 - 2 | 295 | 116 | |
AY819712.1 | - | 1 - 2 | 129 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
