Assay Details
Gene Symbol: LOC5580118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843466.1 | XP_021699158.1 | 1 - 2 | 644 | 61 | |
XM_021843473.1 | XP_021699165.1 | 1 - 2 | 644 | 61 | ||
XM_021843478.1 | XP_021699170.1 | 1 - 2 | 644 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
