Assay Details
Gene Symbol: LOC5572646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654095.2 | XP_001654145.2 | 3 - 4 | 3718 | 73 | |
XM_021845115.1 | XP_021700807.1 | 4 - 5 | 3847 | 73 | ||
GenBank mRNA | AY433562.1 | - | 1 - 2 | 356 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
