Assay Details
Gene Symbol: LOC5573615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660898.2 | XP_001660948.1 | 1 - 2 | 328 | 77 | |
GenBank mRNA | AY433655.1 | - | 1 - 2 | 316 | 77 | |
CR937124.1 | - | 1 - 2 | 311 | 77 | ||
CR937255.1 | - | 1 - 2 | 311 | 77 | ||
CR937831.1 | - | 1 - 2 | 311 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
