Assay Details
Gene Symbol: LOC5566522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650865.2 | XP_001650915.1 | 1 - 2 | 165 | 95 | |
GenBank mRNA | AY433665.1 | - | 1 - 2 | 96 | 95 | |
CR938628.1 | - | 1 - 2 | 97 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001650865.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
