Assay Details
Gene Symbol: LOC5569174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658301.2 | XP_001658351.2 | 1 - 2 | 261 | 69 | |
GenBank mRNA | AY433681.1 | - | 1 - 2 | 190 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
