Assay Details
Gene Symbol: LOC5572081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660113.2 | XP_001660163.1 | 1 - 2 | 261 | 63 | |
GenBank mRNA | AY433709.1 | - | 1 - 2 | 197 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
