Assay Details
Gene Symbol: LOC5579977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658554.2 | XP_001658604.1 | 3 | 1428 | 85 | |
GenBank mRNA | AY433719.1 | - | 1 | 318 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
