Assay Details
Gene Symbol: LOC5572285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653920.2 | XP_001653970.1 | 2 - 3 | 743 | 62 | |
GenBank mRNA | AY433757.1 | - | 2 - 3 | 509 | 62 | |
DQ440263.1 | - | 2 - 3 | 520 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
