Assay Details
Gene Symbol: LOC5570276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658982.2 | XP_001659032.1 | 2 - 3 | 472 | 92 | |
GenBank mRNA | AY445078.1 | - | 2 - 3 | 436 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
