Assay Details
Gene Symbol: LOC5577996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656684.2 | XP_001656734.1 | 2 - 3 | 266 | 63 | |
GenBank mRNA | AY539746.1 | - | 1 - 2 | 41 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
