Assay Details
Gene Symbol: LOC5571497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659697.2 | XP_001659747.2 | 10 - 11 | 6566 | 85 | |
GenBank mRNA | AY713295.1 | - | 10 - 11 | 6530 | 85 |
