Assay Details
Gene Symbol: LOC5571497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659697.2 | XP_001659747.2 | 11 - 12 | 6682 | 106 | |
GenBank mRNA | AY713295.1 | - | 11 - 12 | 6646 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
