Assay Details
Gene Symbol: LOC5571497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659697.2 | XP_001659747.2 | 5 - 6 | 3682 | 65 | |
GenBank mRNA | AY713295.1 | - | 5 - 6 | 3649 | 65 |

More Information
Set Membership: |
![]() ![]() |
