Assay Details
Gene Symbol: LOC5579924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652162.2 | XP_001652212.1 | 1 - 2 | 213 | 103 | |
GenBank mRNA | AY713296.1 | - | 1 - 2 | 201 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001652162.2 transcripts.
Set Membership: |
![]() ![]() |
