Assay Details
Gene Symbol: LOC5579924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652162.2 | XP_001652212.1 | 5 - 6 | 1240 | 84 | |
XM_021850223.1 | XP_021705915.1 | 5 - 6 | 1052 | 84 | ||
GenBank mRNA | AY713296.1 | - | 5 - 6 | 1228 | 84 |
