Assay Details
Gene Symbol: LOC5579996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840597.1 | XP_021696289.1 | 2 - 3 | 1828 | 65 | |
XM_021840598.1 | XP_021696290.1 | 3 - 4 | 1894 | 65 | ||
GenBank mRNA | AY902310.1 | - | 1 - 2 | 224 | 65 | |
AY955097.1 | - | 2 - 3 | 737 | 65 |

More Information
Set Membership: |
![]() ![]() |
