Assay Details
Gene Symbol: LOC5579996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840596.1 | XP_021696288.1 | 3 - 4 | 1626 | 75 | |
XM_021840597.1 | XP_021696289.1 | 3 - 4 | 1870 | 75 | ||
XM_021840598.1 | XP_021696290.1 | 4 - 5 | 1936 | 75 | ||
GenBank mRNA | AY902310.1 | - | 2 - 3 | 266 | 75 | |
AY955097.1 | - | 3 - 4 | 779 | 75 |
