Assay Details
Gene Symbol: LOC5579996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840596.1 | XP_021696288.1 | 8 - 9 | 2633 | 70 | |
XM_021840597.1 | XP_021696289.1 | 8 - 9 | 2877 | 70 | ||
XM_021840598.1 | XP_021696290.1 | 9 - 10 | 2943 | 70 | ||
GenBank mRNA | AY902310.1 | - | 7 - 8 | 1273 | 70 |

More Information
Set Membership: |
![]() ![]() |
