Assay Details
Gene Symbol: LOC5570907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653334.2 | XP_001653384.1 | 4 - 5 | 881 | 114 | |
XM_021844938.1 | XP_021700630.1 | 6 - 7 | 1110 | 114 | ||
XM_021844939.1 | XP_021700631.1 | 4 - 5 | 908 | 114 | ||
XM_021844940.1 | XP_021700632.1 | 4 - 5 | 912 | 114 | ||
GenBank mRNA | AY944756.1 | - | 3 - 4 | 680 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
