Assay Details
Gene Symbol: LOC5570907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653334.2 | XP_001653384.1 | 6 - 7 | 1499 | 140 | |
XM_021844938.1 | XP_021700630.1 | 8 - 9 | 1728 | 140 | ||
XM_021844939.1 | XP_021700631.1 | 6 - 7 | 1526 | 140 | ||
XM_021844940.1 | XP_021700632.1 | 6 - 7 | 1530 | 140 | ||
GenBank mRNA | AY944756.1 | - | 5 - 6 | 1298 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() |
