Assay Details
Gene Symbol: LOC5567962 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847385.1 | XP_021703077.1 | 13 - 14 | 4837 | 79 | |
XM_021847386.1 | XP_021703078.1 | 13 - 14 | 5063 | 79 | ||
XM_021847387.1 | XP_021703079.1 | 13 - 14 | 5059 | 79 | ||
XM_021847388.1 | XP_021703080.1 | 13 - 14 | 5063 | 79 | ||
XM_021847389.1 | XP_021703081.1 | 13 - 14 | 5050 | 79 | ||
XM_021847390.1 | XP_021703082.1 | 13 - 14 | 5057 | 79 | ||
XM_021847391.1 | XP_021703083.1 | 13 - 14 | 5054 | 79 | ||
XM_021847392.1 | XP_021703084.1 | 14 - 15 | 5006 | 79 | ||
XM_021847393.1 | XP_021703085.1 | 13 - 14 | 5057 | 79 | ||
XM_021847394.1 | XP_021703086.1 | 13 - 14 | 5057 | 79 | ||
XM_021847395.1 | XP_021703087.1 | 14 - 15 | 4991 | 79 | ||
XM_021847396.1 | XP_021703088.1 | 14 - 15 | 5006 | 79 | ||
GenBank mRNA | AY971811.1 | - | 10 - 11 | 3558 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
