Assay Details
Gene Symbol: LOC5567962 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847385.1 | XP_021703077.1 | 15 - 16 | 5165 | 103 | |
XM_021847386.1 | XP_021703078.1 | 15 - 16 | 5391 | 103 | ||
XM_021847387.1 | XP_021703079.1 | 15 - 16 | 5387 | 103 | ||
XM_021847388.1 | XP_021703080.1 | 15 - 16 | 5391 | 103 | ||
XM_021847389.1 | XP_021703081.1 | 15 - 16 | 5378 | 103 | ||
XM_021847390.1 | XP_021703082.1 | 15 - 16 | 5385 | 103 | ||
XM_021847391.1 | XP_021703083.1 | 15 - 16 | 5382 | 103 | ||
XM_021847392.1 | XP_021703084.1 | 16 - 17 | 5334 | 103 | ||
XM_021847395.1 | XP_021703087.1 | 16 - 17 | 5319 | 103 | ||
GenBank mRNA | AY971811.1 | - | 12 - 13 | 3886 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
