Assay Details
Gene Symbol: LOC5574977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494668.2 | XP_011492970.2 | 1 - 2 | 147 | 81 | |
GenBank mRNA | CR937128.1 | - | 1 - 2 | 57 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011494668.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
