Assay Details
Gene Symbol: LOC5574977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494668.2 | XP_011492970.2 | 2 - 3 | 364 | 55 | |
GenBank mRNA | CR937128.1 | - | 2 - 3 | 274 | 55 | |
CR938088.1 | - | 1 - 2 | 335 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
