Assay Details
Gene Symbol: LOC5565426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649754.2 | XP_001649804.1 | 3 - 4 | 1249 | 67 | |
GenBank mRNA | CR937211.1 | - | 3 - 4 | 407 | 67 | |
CR937237.1 | - | 3 - 4 | 407 | 67 | ||
CR937259.1 | - | 3 - 4 | 407 | 67 | ||
CR937725.1 | - | 3 - 4 | 407 | 67 | ||
CR937872.1 | - | 3 - 4 | 406 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
