Assay Details
Gene Symbol: LOC5572922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654205.2 | XP_001654255.1 | 1 - 2 | 223 | 130 | |
GenBank mRNA | CR937318.1 | - | 1 - 2 | 188 | 133 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001654205.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
