Assay Details
Gene Symbol: LOC5572922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654205.2 | XP_001654255.1 | 2 - 3 | 532 | 65 | |
GenBank mRNA | CR937318.1 | - | 2 - 3 | 497 | 65 | |
CR937892.1 | - | 2 - 3 | 497 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
