Assay Details
Gene Symbol: LOC5575604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662036.3 | XP_001662086.2 | 2 - 3 | 524 | 93 | |
XM_021838085.1 | XP_021693777.1 | 3 - 4 | 802 | 93 | ||
GenBank mRNA | CR937481.1 | - | 2 - 3 | 529 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
