Assay Details
Gene Symbol: LOC5569203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658324.2 | XP_001658374.1 | 1 | 95 | ||
GenBank mRNA | CR937940.1 | - | 1 | 282 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
