Assay Details
Gene Symbol: LOC5574866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655340.2 | XP_001655390.1 | 2 - 3 | 174 | 76 | |
XM_001655341.2 | XP_001655391.1 | 2 - 3 | 139 | 76 | ||
XM_021853790.1 | XP_021709482.1 | 2 - 3 | 128 | 76 | ||
GenBank mRNA | CR938081.1 | - | 2 - 3 | 111 | 76 | |
CR938529.1 | - | 2 - 3 | 110 | 76 | ||
DQ440229.1 | - | 1 - 2 | 80 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
