Assay Details
Gene Symbol: LOC5576042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662324.2 | XP_001662374.1 | 1 - 2 | 206 | 72 | |
GenBank mRNA | CR938384.1 | - | 1 - 2 | 161 | 72 | |
CR938498.1 | - | 1 - 2 | 147 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001662324.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
