Assay Details
Gene Symbol: LOC5576042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662324.2 | XP_001662374.1 | 4 - 5 | 713 | 86 | |
GenBank mRNA | CR938384.1 | - | 4 - 5 | 668 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
