Assay Details
Gene Symbol: LOC5567888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651873.2 | XP_001651923.1 | 2 - 3 | 402 | 67 | |
GenBank mRNA | CR938479.1 | - | 2 - 3 | 384 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
