Assay Details
Gene Symbol: LOC5580247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662594.2 | XP_001662644.1 | 2 - 3 | 659 | 89 | |
GenBank mRNA | DQ075690.1 | - | 2 - 3 | 610 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
