Assay Details
Gene Symbol: LOC5569705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658646.2 | XP_001658696.2 | 1 - 2 | 242 | 56 | |
GenBank mRNA | DQ177444.1 | - | 1 - 2 | 223 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
