Assay Details
Gene Symbol: LOC5570638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659187.2 | XP_001659237.1 | 1 - 2 | 2052 | 81 | |
GenBank mRNA | DQ342047.1 | - | 1 - 2 | 2052 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
