Assay Details
Gene Symbol: LOC5575914 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655856.2 | XP_001655906.1 | 3 - 4 | 810 | 69 | |
GenBank mRNA | DQ440037.1 | - | 3 - 4 | 647 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
