Assay Details
Gene Symbol: LOC5577889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663539.2 | XP_001663589.2 | 2 - 3 | 390 | 89 | |
XM_001663540.2 | XP_001663590.1 | 2 - 3 | 385 | 89 | ||
GenBank mRNA | DQ440073.1 | - | 2 - 3 | 183 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
