Assay Details
Gene Symbol: LOC5574137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661156.2 | XP_001661206.1 | 3 - 4 | 885 | 99 | |
XM_021851631.1 | XP_021707323.1 | 3 - 4 | 884 | 99 | ||
XM_021851632.1 | XP_021707324.1 | 3 - 4 | 725 | 99 | ||
GenBank mRNA | DQ440127.1 | - | 2 - 3 | 627 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
