Assay Details
Gene Symbol: LOC5567341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651721.2 | XP_001651771.2 | 2 | 475 | 96 | |
GenBank mRNA | DQ440176.1 | - | 1 | 118 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
