Assay Details
Gene Symbol: LOC5571847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653747.3 | XP_001653797.2 | 1 - 2 | 515 | 75 | |
GenBank mRNA | DQ440207.1 | - | 1 - 2 | 185 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
