Assay Details
Gene Symbol: LOC5569076 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658256.2 | XP_001658306.1 | 3 - 4 | 743 | 79 | |
GenBank mRNA | DQ440219.1 | - | 2 - 3 | 607 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
