Assay Details
Gene Symbol: LOC5569076 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658256.2 | XP_001658306.1 | 5 - 6 | 991 | 108 | |
GenBank mRNA | DQ440219.1 | - | 4 - 5 | 855 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
