Assay Details
Gene Symbol: LOC5576852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656230.2 | XP_001656280.2 | 6 - 7 | 1096 | 63 | |
XM_021841836.1 | XP_021697528.1 | 6 - 7 | 871 | 63 | ||
XM_021841837.1 | XP_021697529.1 | 5 - 6 | 628 | 63 | ||
XM_021841838.1 | XP_021697530.1 | 5 - 6 | 826 | 63 |

More Information
Set Membership: |
![]() ![]() |
