Assay Details
Gene Symbol: LOC5576934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656246.2 | XP_001656296.1 | 2 - 3 | 465 | 96 | |
GenBank mRNA | DQ440312.1 | - | 2 - 3 | 177 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
