Assay Details
Gene Symbol: LOC5572148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660175.2 | XP_001660225.2 | 1 - 2 | 333 | 110 | |
GenBank mRNA | AY432879.1 | - | 1 - 2 | 244 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
